missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNAJB12 (aa 270-330) Control Fragment Recombinant Protein

Product Code. 30202601
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202601

Brand: Invitrogen™ RP109303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DNAJB12 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NXW2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54788
Name Human DNAJB12 (aa 270-330) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias DJ10; DKFZp586B2023; DnaJ (Hsp40) homolog, subfamily B, member 12; DnaJ heat shock protein family (Hsp40) member B12; DnaJ homolog 10; dnaJ homolog subfamily B member 12; DnaJ subfamily B member 12-like protein; Dnajb12; LOW QUALITY PROTEIN: dnaJ homolog subfamily B member 12; mDj10
Common Name DNAJB12
Gene Symbol DNAJB12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PPYSLSPRPSVGHIHRRVTDHLGVVYYVGDTFSEEYTGSSLKTVERNVEDDYIANLRNNCW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.