missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNAJB11 (aa 79-210) Control Fragment Recombinant Protein

Product Code. 30197719
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197719

Brand: Invitrogen™ RP90017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53539 (PA5-53539. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DNAJB11 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity bystimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus a glycine/phenylalanine (G/F)-rich region and a C-terminal cysteine-rich region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBS4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51726
Name Human DNAJB11 (aa 79-210) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810031F23Rik; ABBP2; ABBP-2; AL024055; APOBEC1-binding protein 2; cb954; Dj9; Dj-9; DnaJ (Hsp40) homolog, subfamily B, member 11; DnaJ heat shock protein family (Hsp40) member B11; dnaJ homolog subfamily B member 11; DnaJ protein 9; DnaJ protein homolog 9; DNAJB11; dnajb11 protein; DnaJ-like protein subfamily B member 11-like protein; EDJ; Endoplasmic reticulum DNA J domain-containing protein 3; ER-associated DNAJ; ER-associated DNAJ protein 3; ER-associated Hsp40 co-chaperone; ERdj3; ERJ3; ERj3p; ER-resident protein ERdj3; HDJ9; hDj-9; HEDJ; Human DnaJ protein 9; Liver regeneration-related protein LRRGT00084; LRRGT00084; PRO1080; PSEC0121; PWP1-interacting protein 4; UNQ537; UNQ537/PRO1080
Common Name DNAJB11
Gene Symbol DNAJB11
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DSEKRKQYDTYGEEGLKDGHQSSHGDIFSHFFGDFGFMFGGTPRQQDRNIPRGSDIIVDLEVTLEEVYAGNFVEVVRNKPVARQAPGKRKCNCRQEMRTTQLGPGRFQMTQEVVCDECPNVKLVNEERTLEV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.