missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DNA-PKcs Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3415
This item is not returnable.
View return policy
Description
DNA-PKcs Polyclonal specifically detects DNA-PKcs in Human samples. It is validated for Western Blot.
Specifications
| DNA-PKcs | |
| Polyclonal | |
| Unconjugated | |
| P78527 | |
| PRKDC | |
| E. coli-derived recombinant human DNA-PK CS Thr3844-Trp4127 Accession # P78527 | |
| 100 μg | |
| Primary | |
| Detects human DNA-PK CS in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNAPK, DNA-PK catalytic subunit, DNAPKcs, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p350, p460, PRKDC, XRCC7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5591 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction