missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DMPK Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6967
This item is not returnable.
View return policy
Description
DMPK Monoclonal specifically detects DMPK in Human samples. It is validated for Western Blot.
Specifications
| DMPK | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q09013 | |
| DMPK | |
| E. coli-derived recombinant human DMPK Glu393-Thr543 Accession # Q09013 | |
| 100 μg | |
| Primary | |
| Detects human DMPK in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 713931 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DM1, DM1PK, DMK, DMKEC 2.7.11.1, DM-kinase, DMmyotonin protein kinase A, dystrophia myotonica-protein kinase, MDPK, MDPKDM protein kinase, MT-PK, MT-PKmyotonic dystrophy associated protein kinase, Myotonic dystrophy protein kinase, myotonin-protein kinase, thymopoietin homolog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1760 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction