missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DLX5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6710
This item is not returnable.
View return policy
Description
DLX5 Monoclonal specifically detects DLX5 in Human samples. It is validated for Immunocytochemistry.
Specifications
| DLX5 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| P56178 | |
| DLX5 | |
| E. coli-derived recombinant human DLX5 Asn216-Tyr289 Accession # P56178 | |
| 100 μg | |
| Primary | |
| Detects human DLX5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DLX4 is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 712216 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| distal-less homeo box 5, distal-less homeobox 5, homeobox protein DLX-5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1749 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction