missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DLL4 Fc Chimera Recombinant Protein
Description
Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
Specifications
Specifications
| Accession Number | Q9NR61 |
| For Use With (Application) | Control, ELISA Standard, Binding Assay, Western Blot |
| Formulation | PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4 |
| Gene ID (Entrez) | 54567 |
| Molecular Weight (g/mol) | 81 kDa |
| Name | Human DLL4 Fc Chimera |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 50 μg |
| Source | HEK293 cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?