missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DLL4 Fc Chimera Recombinant Protein

Product Code. 17194183
Change view
Click to view available options
Quantity:
5 μg
5 x 5 μg
5 x 50 μg
50 μg
Unit Size:
50µg
5µg
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17194183 5 μg 5µg
17184183 5 x 50 μg Pack of 5
17174183 5 x 5 μg Pack of 5
17104193 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17194183 Supplier Invitrogen™ Supplier No. A42512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NR61
For Use With (Application) Control, ELISA Standard, Binding Assay, Western Blot
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 54567
Molecular Weight (g/mol) 81 kDa
Name Human DLL4 Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 5 μg
Source HEK293 cells
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias AOS6; delta 4; delta ligand 4; delta like canonical Notch ligand 4; Delta4; delta-like 4 (Drosophila); delta-like 4 homolog; delta-like 4 protein; delta-like protein 4; DLL4; Drosophila Delta homolog 4; hdelta2; MGC126344; notch ligand delta-2; notch ligand DLL4; UNQ1895/PRO4341
Common Name DLL4
Gene Symbol Dll4
Biological Activity 1. Measured by its binding ability in a functional ELISA. Immobilized Mouse NOTCH1-His (at 2 μg/mL (100 μL/well) can bind Human DLL4 Fc Chimera Recombinant Protein, the EC50 of human DLL4-hFc is 15-90 ng/mL 2. Measured by the ability of the immobilized protein to enhance BMP2-induced alkaline phosphatase activity in C3H10T1/2 mouse embryonic fibroblast cells. The ED50 for this effect is typically 1-8 μg/mL in the presence of 500 ng/mL recombinant human BMP2.)
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human DLL4, amino acids Met1-Pro524 (Accession # NP_061947.1) with a C-terminal human IgG1 Fc-tag
Content And Storage Includes 1 vial of lyophilized protein. Store at -20 °C.
Activity 1. Measured by its binding ability in a functional ELISA. Immobilized Mouse NOTCH1-His at 2 μg/mL (100 μL/well) can bind Human DLL4 Fc Chimera Recombinant Protein, the EC50 of human DLL4-hFc is 15-90 ng/mL. 2. Measured by the ability of the immobilized protein to enhance BMP2-induced alkaline phosphatase activity in C3H10T1/2 mouse embryonic fibroblast cells. The ED50 for this effect is typically 1-8 μg/mL in the presence of 500 ng/mL recombinant human BMP2.
Endotoxin Level < 1 EU/μg
Expression System HEK293 cells
Protein Family Other Proteins
Form Lyophilized
Protein Subtype Neurotrophic Factors
Research Category Angiogenesis, Drug Development, Oncology, Signal Transduction
Purity or Quality Grade ≥95 % as determined by SDS-PAGE. ≥85 % as determined by SEC-HPLC.
Protein delta-like 4 (Drosophila)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.