missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DLL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£167.00 - £1775.00
Specifications
| Antigen | DLL1 |
|---|---|
| Clone | 251127 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Flow Cytometry, CyTOF, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781187
![]() |
R&D Systems
MAB1818-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743348
![]() |
R&D Systems
MAB1818-100 |
100 μg |
£448.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740927
![]() |
R&D Systems
MAB1818-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| DLL1 | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Delta, delta (Drosophila)-like 1, Delta 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| DLL1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 251127 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| AAG09716 | |
| 28514 | |
| Mouse myeloma cell line NS0-derived recombinant human DLL1 Ser22-Gly540 Accession # AAG09716 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant human DLL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse DLL4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title