missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DLEC/CLEC4C/BDCA-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1376-SP
This item is not returnable.
View return policy
Description
DLEC/CLEC4C/BDCA-2 Polyclonal specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| DLEC/CLEC4C/BDCA-2 | |
| Polyclonal | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
| Q8WTT0 | |
| CLEC4C | |
| Mouse myeloma cell line NS0-derived recombinant human DLEC/CLEC4C/BDCA-2 Phe46-Ile213 Accession # Q8WTT0 | |
| 25 μg | |
| Primary | |
| Detects human DLEC/CLEC4C/BDCA-2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLEC4C, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 170482 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction