missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-3 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB1118-500
Additional Details : Weight : 0.00970kg
Description
Dkk-3 Monoclonal specifically detects Dkk-3 in Human samples. It is validated for Western Blot.Specifications
Dkk-3 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9UBP4.1 | |
DKK3 | |
Mouse myeloma cell line NS0-derived recombinant human Dkk-3 Ala22-Ile350 Accession # Q9UBP4.1 | |
500 μg | |
Primary | |
Detects human Dkk-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Dkk-1, rmDkk-3, or rhDkk-4. | |
Human | |
Purified |
Western Blot | |
138726 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk3, Dkk-3, hDkk-3, regulated in glioma, REIC, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27122 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |