missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human DKK-1 Recombinant Protein, PeproTech®

Product Code. 17870393
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17870393

Brand: Gibco™ 12030500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Mature human DKK-1 expressed in HEK293 cells is a 35-40 kDa glycoprotein containing 235 amino acid residues. The calculated molecular weight of Recombinant Human DKK-1 expressed in HEK293 cells is 25.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O94907
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 22943
Molecular Weight (g/mol) 25.8 kDa
Name Human DKK-1
Quantity 500 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias dickkopf 1 homolog; dickkopf homolog 1; dickkopf homolog 1 (Xenopus laevis); dickkopf related protein-1; dickkopf WNT signaling pathway inhibitor 1; Dickkopf1; dickkopf-1; dickkopf-1 like; dickkopf-like protein 1; dickkopf-related protein 1; DKK1; DKK-1; hDkk1; hDkk-1; mdkk-1; SK; UNQ492/PRO1008
Common Name DKK1
Gene Symbol DKK1
Biological Activity Determined by its ability to inhibit mWnt3a-induced TCF/LEF-luciferase activity in reporter HEK293 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence TLNSVLNSNA IKNLPPPLGG AAGHPGSAVS AAPGILYPGG NKYQTIDNYQ PYPCAEDEEC GTDEYCASPT RGGDAGVQIC LACRKRRKRC MRHAMCCPGN YCKNGICVSS DQNHFRGEIE ETITESFGND HSTLDGYSRR TTLSSKMYHT KGQEGSVCLR SSDCASGLCC ARHFWSKICK PVLKEGQVCT KHRRKGSHGL EIFQRCYCGE GLSCRIQKDH HQASNSSRLH TCQRH
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.