missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Dkk-1 Recombinant Protein
Recombinant Human Dickkopf Homolog 1 (DKK1) is a bioactive protein intended for use in cell culture applications
£391.00 - £7870.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 42 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | DKK-1 |
Biological Activity | Linear range of 0.3 to 40 ng⁄ml |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10023534
|
Gibco™
PHC9214 |
10μg |
£391.00
Each |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115114
|
Gibco™
PHC9215 |
25μg |
£924.00
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10227904
|
Gibco™
PHC9211 |
100μg |
£2760.00
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10758225
|
Gibco™
PHC9213 |
1mg |
£7870.00
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human Dkk-1 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
DKK1 is a member of the dickkopf family. It is a secreted protein with two cysteine-rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by a functional ELISA measuring the ability of human DKK1 to bind immobilized recombinant human LRP6/Fc chimera receptor
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
Linear range of 0.3 to 40 ng⁄ml | |
Store lyophilized human DKK1 at 2 to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤ -20°C. Avoid repeated freeze⁄thaw cycles. | |
< 0.1 ng/μg | |
Wet Ice | |
Mammalian | |
Recombinant, Ligand | |
Other Proteins | |
Gibco™ |
42 kDa | |
DKK-1 | |
Human | |
Carrier-Free | |
Human | |
Linear range of 0.3 to 40 ng⁄ml | |
Cytokines & Receptors | |
Lyophilized | |
Oncology, Signal Transduction | |
>95% by SDS-PAGE |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.