missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1096-500
Additional Details : Weight : 0.00970kg
Description
Dkk-1 Monoclonal specifically detects Dkk-1 in Human samples. It is validated for Western Blot.Specifications
Dkk-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O94907 | |
DKK1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907 | |
500 μg | |
Primary | |
Detects human Dkk-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 50% cross-reactivity with recombinant mouse (rm) Dkk-1 and no cross-reactivity with rmDkk-2, rhDkk-3, or rhDkk-4. | |
Human | |
Purified |
Western Blot | |
141119 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
22943 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |