missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DISP2 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Disp2 Synthetic Peptide, PEP-0751, from Invitrogen.
DISP2, also known as dispatched homolog 2, is a protein that plays a critical role in the regulation of the hedgehog signaling pathway. It is involved in the transport and release of hedgehog ligands, which are important for cellular differentiation and tissue development. Mutations in the DISP2 gene have been associated with various developmental disorders, including holoprosencephaly and cleft lip and palate. DISP2 is also being studied as a potential therapeutic target in cancer, as the hedgehog pathway is frequently dysregulated in various types of tumors
Specifications
Specifications
| Gene ID (Entrez) | 85455 |
| Species | Human |
| Purification Method | purified |
| Accession Number | A7MBM2 |
| Common Name | Human DISP2 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction