missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DIDO1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6947
This item is not returnable.
View return policy
Description
DIDO1 Polyclonal specifically detects DIDO1 in Human samples. It is validated for Western Blot.
Specifications
| DIDO1 | |
| Polyclonal | |
| Unconjugated | |
| Q9BTC0 | |
| DIDO1 | |
| E. coli-derived recombinant human DIDO1 Met1-Ser117 Accession # Q9BTC0 | |
| 100 μg | |
| Primary | |
| Detects human DIDO1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| BYE1, C20orf158, DATF1, DATF-1, DATF1DIDO3, death associated transcription factor 1, death inducer-obliterator 1, Death-associated transcription factor 1, death-inducer obliterator 1, DIO1, DIO1DIDO2, DIO-1hDido1, dJ885L7.8, DKFZp434P1115, FLJ11265, KIAA0333chromosome 20 open reading frame 158, MGC16140 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 11083 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction