missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DGK-delta Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7426
This item is not returnable.
View return policy
Description
DGK-delta Monoclonal specifically detects DGK-delta in Human samples. It is validated for Western Blot.
Specifications
| DGK-delta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q16760 | |
| DGKD | |
| E. coli-derived recombinant human DGF-delta Ile570-Thr717 Accession # Q16760 | |
| 100 μg | |
| Primary | |
| Detects human DGF-delta in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 625423 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 130 kDa diacylglycerol kinase, DAG kinase delta, DGKD, dgkd-2, DGKdelta, DGK-delta, diacylglycerol kinase delta, diacylglycerol kinase, delta (130kD), diacylglycerol kinase, delta 130kDa, Diglyceride kinase delta, FLJ26930, KIAA0145EC 2.7.1.107 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8527 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction