missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human DGAT1 (NP_036211) VersaClone cDNA

Product Code. 18757658
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18757658

Brand: R&D Systems™ RDC2928

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene DGAT1
Accession Number NP_036211
Gene ID (Entrez) 8694
Gene Alias ACAT-related gene product 1, AGRP1, ARAT, ARGP1, ARGP1acyl coenzyme A:cholesterol acyltransferase related gene 1, DGAT, DGATACAT related gene product 1, diacylglycerol O-acyltransferase 1, diacylglycerol O-acyltransferase homolog 1, diacylglycerol O-acyltransferase homolog 1 (mouse), DIAR7, Diglyceride acyltransferase, EC 2.3.1, EC 2.3.1.20
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.