missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Desmoglein-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6589
This item is not returnable.
View return policy
Description
Desmoglein-4 Monoclonal specifically detects Desmoglein-4 in Human samples. It is validated for Western Blot.
Specifications
| Desmoglein-4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q86SJ6 | |
| DSG4 | |
| Mouse myeloma cell line NS0-derived recombinant human Desmoglein-4 Glu24-Ala631 Accession # Q86SJ6 | |
| 100 μg | |
| Primary | |
| Detects human Desmoglein-4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Desmoglein-1, -2, or -3 is observed and approximately 40% cross-reactivity with recombinant mouse Desmoglein-4 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 683135 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cad13, Cadherin family member 13, CDGF13, CDHF13, CDHF13CDH family member 13, desmoglein 4, Desmoglein4, DSG4, LAHdesmoglein-4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 147409 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction