missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Desmoglein-1 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF944
This item is not returnable.
View return policy
Description
Desmoglein-1 Polyclonal specifically detects Desmoglein-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Desmoglein-1 | |
| Polyclonal | |
| Biotin | |
| Q02413 | |
| DSG1 | |
| Mouse myeloma cell line NS0-derived recombinant human Desmoglein-1 Propeptide Met1-His545 Accession # Q02413 | |
| 50 μg | |
| Primary | |
| Detects human Desmoglein-1 Propeptide in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Desmoglein-2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Cadherin family member 4, CDHF4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmoglein1, Desmosomal glycoprotein 1, DG1, DGI, DSG, DSG1, Pemphigus foliaceus antigen, SPPK1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1828 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction