Learn More
Human Desmocollin-1 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB4955-100
 View more versions of this product
Product Code. 18732007
  • £581.00 / 100µg
Description
Desmocollin-1 Monoclonal antibody specifically detects Desmocollin-1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Desmocollin-1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, Desmocollin1, desmosomal glycoprotein 2/3, DG2/DG3, DSC1
Mouse myeloma cell line, NS0-derived human Desmocollin-1, Arg135-Asn686, Accession # Q08554
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG
Immunohistochemistry
2906A
Immunohistochemistry 3-25 ug/mL
Q08554
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
1823
Human
Purified