missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DEPDC6 (aa 210-309) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Negative regulator of the mTORC1 and mTORC2 signaling pathways. Inhibits the kinase activity of both complexes.
Specifications
Specifications
| Accession Number | Q8TB45 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 64798 |
| Name | Human DEPDC6 (aa 210-309) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | DEP domain containing 6; DEP domain containing MTOR interacting protein; DEP domain containing MTOR-interacting protein; DEP domain-containing mTOR-interacting protein; DEP domain-containing protein 6; DEP0.6; Depdc6; DEPTOR; Kiaa4200; RGD1561030 |
| Common Name | DEPDC6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction