missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DENND6B (aa 347-425) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53880 (PA5-53880. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q8NEG7 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 414918 |
| Name | Human DENND6B (aa 347-425) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700027J05Rik; AFI1B; AI414881; DENN domain containing 6 B; DENN domain-containing protein 6 B; DENN/MADD domain containing 6 B; Dennd6b; FAM116B; family with sequence similarity 116, member B; hypothetical protein MGC38041; protein DENND6B; protein FAM116B; RGD1308012 |
| Common Name | DENND6B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu