missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Dematin (aa 285-360) Control Fragment Recombinant Protein

Product Code. 30205722
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205722

Brand: Invitrogen™ RP93854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55040 (PA5-55040. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is an actin binding and bundling protein that plays a structural role in erythrocytes, by stabilizing and attaching the spectrin/actin cytoskeleton to the erythrocyte membrane in a phosphorylation-dependent manner. This protein contains a core domain in the N-terminus, and a headpiece domain in the C-terminus that binds F-actin. When purified from erythrocytes, this protein exists as a trimer composed of two 48 kDa polypeptides and a 52 kDa polypeptide. The different subunits arise from alternative splicing in the 3' coding region, where the headpiece domain is located. Disruption of this gene has been correlated with the autosomal dominant Marie Unna hereditary hypotrichosis disease, while loss of heterozygosity of this gene is thought to play a role in prostate cancer progression. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08495
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2039
Name Human Dematin (aa 285-360) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI325486; dematin; dematin actin binding protein; dematin actin-binding protein; DMT; Dmtn; Epb4.9; Epb49; Erythrocyte membrane protein band 4.9; erythrocyte membrane protein band 4.9 (dematin); erythrocyte protein band 4.9
Common Name Dematin
Gene Symbol DMTN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SKSSSLPAYGRTTLSRLQSTEFSPSGSETGSPGLQNGEGQRGRMDRGNSLPCVLEQKIYPYEMLVVTNKGRTKLPP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.