missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Decorin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB143-100
This item is not returnable.
View return policy
Description
Decorin Monoclonal specifically detects Decorin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Decorin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Simple Western 10 μg/mL | |
| NP_598013.1 | |
| DCN | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Decorin Gly17-Lys172 Accession # NP_598013.1 | |
| 100 μg | |
| Primary | |
| Detects human Decorin in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse Decorin. | |
| Human | |
| Supernatant |
| Western Blot, Immunohistochemistry | |
| 115402 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bone proteoglycan II, CSCD, DCN, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-II, PGS2, PG-S2, proteoglycan core protein, SLRR1B, SLRR1BPGS2, small leucine-rich protein 1B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1634 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction