missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Decorin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 18 publications
Brand: R&D Systems AF143-SP
This item is not returnable.
View return policy
Description
Decorin Polyclonal specifically detects Decorin in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Decorin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bone proteoglycan II, CSCD, DCN, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-II, PGS2, PG-S2, proteoglycan core protein, SLRR1B, SLRR1BPGS2, small leucine-rich protein 1B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1634 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 10-20 ug/mL | |
| P07585 | |
| DCN | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Decorin Gly17-Lys359 Accession # P07585 | |
| 25 μg | |
| Primary | |
| Detects human Decorin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction