missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DDX42 (aa 39-150) Control Fragment Recombinant Protein

Product Code. 30203596
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203596

Brand: Invitrogen™ RP93545

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54828 (PA5-54828. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the Asp-Glu-Ala-Asp (DEAD) box protein family. Members of this protein family are putative RNA helicases, and are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Two transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86XP3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11325
Name Human DDX42 (aa 39-150) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810047H21Rik; ATP-dependent RNA helicase DD x 42; AW319508; AW556242; B430002H05Rik; Dd x 42; DD x 42 P; DEAD (Asp-Glu-Ala-Asp) box helicase 42; DEAD (Asp-Glu-Ala-Asp) box polypeptide 42; DEAD box protein 42; DEAD-box helicase 42; RHELP; RNA helicase-like protein; RNA helicase-related protein; RNAHP; SF3b DEAD box protein; SF3b125; SF3B8; splicing factor 3 b, subunit 8; splicing factor 3 B-associated 125 kDa protein
Common Name DDX42
Gene Symbol DDX42
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FGATSSSSGFGKSAPPQLPSFYKIGSKRANFDEENAYFEDEEEDSSNVDLPYIPAENSPTRQQFHSKPVDSDSDDDPLEAFMAEVEDQAARDMKRLEEKDKERKNVKGIRDD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.