missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DDX26B (aa 708-798) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65078 (PA5-65078. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DDX26B is a protein coding gene. An important paralog of this gene is INTS6.
Specifications
Specifications
| Accession Number | Q5JSJ4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 203522 |
| Name | Human DDX26B (aa 708-798) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4930535D10Rik; 6330505F04Rik; D130066O12; dd x 26 b; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26 B; hypothetical protein MGC63527; int6; Integrator complex subunit 6; integrator complex subunit 6 like; integrator complex subunit 6-like; ints6; Ints6l; LOW QUALITY PROTEIN: integrator complex subunit 6-like; protein DD x 26 B; zgc:63527 |
| Common Name | DDX26B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction