missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DDR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2396-SP
This item is not returnable.
View return policy
Description
DDR1 Monoclonal specifically detects DDR1 in Human samples. It is validated for Immunoprecipitation.
Specifications
| DDR1 | |
| Monoclonal | |
| LYOPH | |
| Immunoprecipitation 25 ug/mL | |
| Q08345 | |
| DDR1 | |
| Mouse myeloma cell line NS0-derived recombinant human DDR1 Asp19-Thr416 Accession # Q08345 | |
| 25 μg | |
| Primary | |
| Detects human DDR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human DDR2. | |
| Human | |
| Purified |
| Immunoprecipitation | |
| 290420 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAK, CD167 antigen-like family member A, CD167a, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, RTK6, TRK E, TrkE | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 780 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction