missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DDI1 (aa 362-394) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (56%), Rat (56%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140306 (PA5-140306. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
Specifications
Specifications
| Accession Number | Q8WTU0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 414301 |
| Name | Human DDI1 (aa 362-394) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700011N24Rik; DDI1; DDI1, DNA-damage inducible 1, homolog 1; DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae); DNA damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1 (S. cerevisiae); DNA-damage inducible protein 1; protein DDI1 homolog 1; RGD1559430 |
| Common Name | DDI1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction