missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DCUN1D2 (aa 14-72) Control Fragment Recombinant Protein

Product Code. 30180583
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180583

Brand: Invitrogen™ RP97975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58748 (PA5-58748. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Potently stimulates the neddylation of cullin components of SCF-type E3 ubiquitin ligase complexes from the NEDD8-conjugating E2 enzyme UBC12. Neddylation of cullins play an essential role in the regulation of SCF-type complexes activity. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6PH85
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55208
Name Human DCUN1D2 (aa 14-72) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW495713; C13orf17; DCN1, defective in cullin neddylation 1, domain containing 2; DCN1, defective in cullin neddylation 1, domain containing 2 (S. cerevisiae); DCN1-like protein 2; DCN1-like protein 2; LOW QUALITY PROTEIN: DCN1-like protein 2; DCUN1 domain-containing protein 2; Dcun1d2; DCUN1L2; defective in cullin neddylation 1 domain containing 2; defective in cullin neddylation protein 1-like protein 2; mFLJ10704; RGD1311743; squamous cell carcinoma-related oncogene
Common Name DCUN1D2
Gene Symbol DCUN1D2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QFMACTQAGERTAIYCLTQNEWRLDEATDSFFQNPDSLHRESMRNAVDKKKLERLYGRY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.