missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DcR3/TNFRSF6B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB21351
This item is not returnable.
View return policy
Description
DcR3/TNFRSF6B Monoclonal specifically detects DcR3/TNFRSF6B in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| DcR3/TNFRSF6B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O95407 | |
| TNFRSF6B | |
| Mouse myeloma cell line NS0-derived recombinant human DcR3/TNFRSF6B Val24-His300 Accession # O95407 | |
| 500 μg | |
| Primary | |
| Detects human DcR3/TNFRSF6B in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) OPG, recombinant mouse (rm) OX40, rhRANK, rhRELT, rhTAJ, rhTNF RI, rhTNF RII, rhTRADD, rhTWEAK R, rhTRAIL RI, rhTRAIL R2, rhTRAIL R3, or rhTRAIL R4 is observed. | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 229817 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68, M68DJ583P15.1.1, TNFRSF6B, TR6, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8771 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction