missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DCP1B (aa 132-236) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58894 (PA5-58894. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.
Specifications
Specifications
| Accession Number | Q8IZD4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 196513 |
| Name | Human DCP1B (aa 132-236) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | B930050E02Rik; DCP1; DCP1 decapping enzyme homolog b; DCP1 decapping enzyme homolog B (S. cerevisiae); Dcp1b; decapping enzyme hDcp1b; decapping mRNA 1 B; hDcp1b; mRNA-decapping enzyme 1 B; RGD1562214 |
| Common Name | DCP1B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction