missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DCLK3 (aa 207-321) Control Fragment Recombinant Protein

Product Code. 30205650
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205650

Brand: Invitrogen™ RP109973

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (53%), Rat (53%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144847 (PA5-144847. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DCLK3 is one of three doublecortin-like kinases similar to the Ca2+/calmodulin-dependent protein kinase (CaMK) family. DCLK3 mRNA, like that of the homologous DCLK1 and DCLK3, is highly expressed in adult brain, but only DCLK3 transcripts are present in liver and kidney, suggesting that DCLK3 may play other roles than in cortical development. The DCLK proteins are homologous to Doublecortin (DCX), a protein that is mutated in X-linked human lissencephaly. In mouse models where the DCX gene has been disrupted, DCLK1 expression increases slightly and appears to compensate for the loss of DCX, as mice mutant for both DCX and DCLK1 show a severe phenotype including perinatal lethality, disorganized neocortical layering, and profound hippocampal cytoarchitectural disorganization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9C098
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 85443
Name Human DCLK3 (aa 207-321) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BC056929; C730036H08; CLICK-I and II-related; Click-I, -II related; CLICK-I,II-related; CLR; Dcamkl3; DCDC3C; DCK3; Dclk3; doublecortin and CaM kinase-like 3; doublecortin domain-containing protein 3 C; doublecortin like kinase 3; doublecortin-like and CAM kinase-like 3; Doublecortin-like kinase 3; hCG_16695; KIAA1765; RGD1309232; serine/threonine-protein kinase DCLK3
Common Name DCLK3
Gene Symbol DCLK3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PRNPTQELRRPSKSMDKKEDRGPEDQESHAQGAAKAKKDLVEVLPVTEEGLREVKKDTRPMSRSKHGGWLLREHQAGFEKLRRTRGEEKEAEKEKKPCMSGGRRMTLRDDQPAKL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.