missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DCIR/CLEC4A Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1748-SP
This item is not returnable.
View return policy
Description
DCIR/CLEC4A Polyclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| DCIR/CLEC4A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD367, CLEC4A, CLECSF6, CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor, LLIR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 50856 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9UMR7 | |
| CLEC4A | |
| Mouse myeloma cell line NS0-derived recombinant human DCIR/CLEC4A isoform 1 Gln70-Leu237 Accession # Q9UMR7 | |
| 25 μg | |
| Primary | |
| Detects human DCIR/CLEC4A in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction