missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DCHS2 (aa 2519-2596) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Förpackningsstorlek:
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64156 (PA5-64156. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spezifikation
Spezifikation
| Accession Number | Q6V1P9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 54798 |
| Name | Human DCHS2 (aa 2519-2596) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | cadherin-27; cadherin-like 27; cadherin-like protein CDHJ; Cadherin-like protein VR8; cadherin-related family member 7; CDH27; CDHJ; CDHR7; dachsous cadherin-related 2; DCHS2; PCDH23; PCDHJ; Protein dachsous homolog 2; protocadherin 23; protocadherin PCDHJ; Protocadherin-23 |
| Common Name | DCHS2 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur