missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DC8 (aa 131-199) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65817 (PA5-65817. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis.
Specifications
Specifications
| Accession Number | Q96IY1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 25936 |
| Name | Human DC8 (aa 131-199) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4833432M17Rik; AI451597; C1orf48; DC31; DC8; Gm105; Gm984; kinetochore-associated protein NSL1 homolog; MIS14; NSL1; NSL1, MIND kinetochore complex component, homolog; NSL1, MIND kinetochore complex component, homolog (S. cerevisiae); NSL1, MIS12 kinetochore complex component; RGD1562052 |
| Common Name | DC8 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction