missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DC-STAMP Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7824U-100UG
This item is not returnable.
View return policy
Description
DC-STAMP/TM7SF4 Monoclonal specifically detects DC-STAMP/TM7SF4 in Human samples. It is validated for Flow Cytometry.
Specifications
| DC-STAMP/TM7SF4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9H295 | |
| DCSTAMP | |
| E. coli-derived recombinant human DC-STAMP Asp314-Thr376 Accession # Q9H295 | |
| RUO | |
| 81501 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 788524 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DCSTAMP, dendritic cell-specific transmembrane protein, FIND, IL-4-induced Protein, TM7SF4, transmembrane 7 superfamily member 4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human DC-STAMP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse DC-STAMP is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction