missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DC-SCRIPT/ZNF366 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF4707-SP
This item is not returnable.
View return policy
Description
DC-SCRIPT/ZNF366 Polyclonal specifically detects DC-SCRIPT/ZNF366 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| DC-SCRIPT/ZNF366 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DCSCRIPT, FLJ39796, zinc finger protein 366, ZNF366 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 167465 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q8N895 | |
| ZNF366 | |
| E. coli-derived recombinant human DC-SCRIPT/ZNF366 Met559-Ile744 Accession # Q8N895 | |
| 25 μg | |
| Primary | |
| Detects human DC-SCRIPT/ZNF366 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) ZNF24 and rhZNF206 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction