missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DBI (aa 1-39) Control Fragment Recombinant Protein

Product Code. 30195957
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195957

Brand: Invitrogen™ RP103098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84066 (PA5-84066. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes diazepam binding inhibitor, a protein that is regulated by hormones and is involved in lipid metabolism and the displacement of beta-carbolines and benzodiazepines, which modulate signal transduction at type A gamma-aminobutyric acid receptors located in brain synapses. The protein is conserved from yeast to mammals, with the most highly conserved domain consisting of seven contiguous residues that constitute the hydrophobic binding site for medium- and long-chain acyl-Coenzyme A esters. Diazepam binding inhibitor is also known to mediate the feedback regulation of pancreatic secretion and the postprandial release of cholecystokinin, in addition to its role as a mediator in corticotropin-dependent adrenal steroidogenesis. Three pseudogenes located on chromosomes 6, 8 and 16 have been identified. Multiple transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07108
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1622
Name Human DBI (aa 1-39) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ACBD 1; ACBD1; ACBP; Acoabp3; acyl coenzyme A binding protein; acyl-CoA binding protein; acyl-CoA-binding protein; acyl-Coenzyme A binding domain containing 1; CCK RP; CCKRP; CCK-RP; cholecystokinin-releasing peptide, trypsin-sensitive; Dbi; diazepam binding inhibitor; Diazepam binding inhibitor (GABA receptor modulator acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein); Diazepam binding inhibitor (GABA receptor modulator, acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-Coenzyme A binding protein); diazepam binding inhibitor, acyl-CoA binding protein; diazepam binding inhibitor, splice form 1 b; diazepam-binding inhibitor; Endozepine; EP; GABA receptor modulator; LRRGT00046; MGC70414; octadecaneuropeptide; ODN; RNACOABP3; Triakontatetraneuropeptide; Ttn; valiμm
Common Name DBI
Gene Symbol DBI
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MSQAEFEKAAEEVRHLKTKPSDEEMLFIYGHYKQATVGD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.