missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAP10/HCST Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9786T-100UG
This item is not returnable.
View return policy
Description
DAP10/HCST Monoclonal specifically detects DAP10/HCST in Human samples. It is validated for Flow Cytometry.
Specifications
| DAP10/HCST | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| DAP10PIK3AP, DKFZP586C1522, HCST, hematopoietic cell signal transducer, KAP10, KAP10DNAX-activation protein 10, kinase assoc pro of ∼10kDa, kinase assoc protein, Membrane protein DAP10, phosphoinositide-3-kinase adaptor protein, PIK3AP, Transmembrane adapter protein KAP10 | |
| Synthetic peptide containing human DAP10/HCST Accession # Q9UBK5 | |
| 100 μg | |
| 10870 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 982129 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9UBK5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human DAP10/HCST in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction