missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytosolic Sulfotransferase 4A1/SULT4A1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5826-SP
This item is not returnable.
View return policy
Description
Produced in goats immunized with purified, E. coli-derived, recombinant human Cytosolic Sulfotransferase 4A1 isoform 1 (rhSULT4A1; 2 - 284; Accession # Q9BR01). Human SULT4A1 specific IgG was purified by human SULT4A1 affinity chromatography.
Cytosolic Sulfotransferase 4A1/SULT4A1 Polyclonal specifically detects Cytosolic Sulfotransferase 4A1/SULT4A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Cytosolic Sulfotransferase 4A1/SULT4A1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brain sulfotransferase-like protein, BRSTL1, BR-STL-1, EC 2.8.2.-, hBR-STL, hBR-STL-1MGC40032, nervous system cytosolic sulfotransferase, Nervous system sulfotransferase, NST, ST4A1, sulfotransferase 4A1, sulfotransferase family 4A, member 1, sulfotransferase-related protein, SULT4A1, SULTX3, SULTX3DJ388M5.3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25830 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9BR01 | |
| SULT4A1 | |
| E. coli-derived recombinant human SULT4A1 isoform 1 Ala2-Leu284 Accession # Q9BR01 | |
| 25 μg | |
| Primary | |
| Detects human SULT4A1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) SULT1A1, rhSULT2A1, and rhSULT1E1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction