missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytosolic Sulfotransferase 1C2/SULT1C2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7458-SP
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1C2/SULT1C2 Polyclonal specifically detects Cytosolic Sulfotransferase 1C2/SULT1C2 in Human samples. It is validated for Western Blot.
Specifications
| Cytosolic Sulfotransferase 1C2/SULT1C2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cytosolic, 1C, member 1, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.2, humSULTC2, ST1C1, ST1C2, Sulfotransferase 1C1, sulfotransferase family, cytosolic, 1C, member 2, SULT1C#1, SULT1C1, SULT1C2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6819 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O00338 | |
| SULT1C2 | |
| E. coli-derived recombinant human Cytosolic Sulfotransferase 1C2/SULT1C2 Ala2-Leu296, predicted Accession # O00338 | |
| 25 μg | |
| Primary | |
| Detects human Cytosolic Sulfotransferase 1C2/SULT1C2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human SULT1C4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction