missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Cytokine A Premixed Magnetic Luminex™ Performance Assay

Product Code. 18422100
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
missing translation for 'unitSize'
1 set
Each
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422100 19 Plex 1 set
18491400 18 Plex 1 set
18481530 21 Plex 1 set
18462120 22 Plex 1 set
18452520 13 Plex 1 set
18431210 20 Plex 1 set
18482300 2 Plex 1 set
18451950 11 Plex 1 set
18432230 17 Plex 1 set
18482650 3 Plex 1 set
18442120 4 Plex 1 set
18482520 14 Plex 1 set
18482450 10 Plex 1 set
18442320 9 Plex 1 set
18401440 7 Plex 1 set
18492220 8 Plex 1 set
18481430 12 Plex 1 set
18451320 6 Plex 1 set
18481920 15 Plex 1 set
18482400 16 Plex 1 set
30001671 5 Plex Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18422100 missing translation for 'mfr' R&D Systems™ missing translation for 'supplierNo' FCSTM0319

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ platform

TRUSTED_SUSTAINABILITY

Specifications

Quantity 19 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.