missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cytokeratin 6A (aa 531-564) Control Fragment Recombinant Protein

Product Code. 30210646
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210646

Brand: Invitrogen™ RP103999

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84483 (PA5-84483. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02538
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3853
Name Human Cytokeratin 6 A (aa 531-564) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310016L08Rik; 60-kDa keratin; CK6A; CK-6 A; CK6C; CK-6 C; CK6D; CK-6 D; CK-6 E; cytokeratin 6 A; cytokeratin 6 C; cytokeratin 6 D; cytokeratin-6 A; cytokeratin-6 C; Cytokeratin-6 D; cytokeratin-6 E; Hom s 5; K6A; K6a keratin; K6C; K6D; kamp-keratin derived antimicrobial peptide; KDAMP; Ker2; keratin 6 A; keratin 6 A, , type II; keratin 6 A, type II; keratin complex 2, basic, gene 6 A; keratin complex 2, basic, gene 6 c; keratin complex 2, gene 6 A; keratin K6; keratin K6h; keratin, epidermal type II, K6A; keratin, type II cytoskeletal 6 A; keratin, type II cytoskeletal 6 C; Keratin-6 A; keratin-6-alpha; keratin-6 C; Krt2-6; Krt2-6 A; Krt2-6 c; Krt6; KRT6A; KRT6C; KRT6D; LOC683313; mK6[a]; MK6a; mK6-alpha; PC3; similar to keratin complex 2, basic, gene 6 A; type-II keratin Kb12; type-II keratin Kb6
Common Name Cytokeratin 6 A
Gene Symbol KRT6A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SGRAIGGGLSSVGGGSSTIKYTTTSSSSRKSYKH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.