missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytokeratin 19 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF3506-SP
This item is not returnable.
View return policy
Description
Cytokeratin 19 Polyclonal specifically detects Cytokeratin 19 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Specifications
| Cytokeratin 19 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, EndoC, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, Krt19, MGC15366 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3880 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P08727 | |
| KRT19 | |
| E. coli-derived recombinant human Cytokeratin 19 Thr2-Leu400 Accession # P08727 | |
| 25 μg | |
| Primary | |
| Detects human Cytokeratin 19 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Cytokeratin 14 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction