missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cytokeratin 13 (aa 410-458) Control Fragment Recombinant Protein

Product Code. 30200564
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200564

Brand: Invitrogen™ RP108558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (37%), Rat (37%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84825 (PA5-84825. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ∽40 kDa and ∽68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13646
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3860
Name Human Cytokeratin 13 (aa 410-458) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 47 kDa cytokeratin; CK13; CK-13; cytokeratin 13; cytokeratin-13; K13; K-13; Ka13; keratin 13; keratin 13, type I; keratin complex 1, acidic, gene 13; keratin, type I cytoskeletal 13; Keratin-13; Krt-1.13; Krt1-13; KRT13; krt13 {ECO:0000250; MGC161462; MGC3781; Type I keratin Ka13; UniProtKB:P13646}; WSN2
Common Name Cytokeratin 13
Gene Symbol KRT13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EGQDAKMIGFPSSAGSVSPRSTSVTTTSSASVTTTSNASGRRTSDVRRP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.