missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cytochrome C ELISA Kit

Product Code. 15561847
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
96 tests
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15561847 10 x 96 Tests Pack of 10
15541847 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15561847 Supplier Invitrogen™ Supplier No. BMS263TEN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cytochrome c
Biotin-Conjugate anti-human Cytochrome c monoclonal antibody
Streptavidin-HRP
Human Cytochrome c Standard lyophilized, 10ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Lysis Buffer 10x
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Green-Dye
Red-Dye
Adhesive Films

ELISA

The Human Cytochrome c ELISA quantitates Hu cytochrome c in human cell lysates. The assay will exclusively recognize both natural and recombinant Hu cytochrome c. Principle of the method The Human cytochrome c solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P99999
Assay Range 0.08 to 5.0 ng/mL
Assay Sensitivity 0.05 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 54205
Gene Symbol CYC,HCS,THC4
Interassay CV 4%
Intraassay CV 6
Kit Contents Pre-coated 96 well plates, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Lysis Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 5 μL
Storage Requirements 2°C to 8°C
Synonym CYC, somatic Cytochrome C, HCS
Target Cytochrome C
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 10 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.