missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Cytochrome c (Human Heart Tissue) Protein
Click to view available options
:
10ug; Carrier Free
Description
The Human Cytochrome c (Human Heart Tissue) Protein is derived from Human Heart. The Human Cytochrome c (Human Heart Tissue) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 54205 |
| Name | Cytochrome c |
| Quantity | 10μg |
| Source | Human heart tissue-derived Cytochrome c protein The human heart tissue used for the isolation of this product was certified by the supplier to be non-reactive for anti-HIV-1/2,anti-HCV,HBsAg,and HIV-1 Ag at the time of shipment. Human products should always be treated in accordance with universal handling precautions. Cytochrome c was chemically oxidized during purification. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction