missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin SA Antibody, R&D Systems™

Mouse Monoclonal Antibody
£147.00 - £452.00
Specifications
| Antigen | Cystatin SA |
|---|---|
| Clone | 202332 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753749
![]() |
R&D Systems
MAB1201-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18113161
![]() |
R&D Systems
MAB1201 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin SA Monoclonal specifically detects Cystatin SA in Human samples. It is validated for Western Blot.Specifications
| Cystatin SA | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P09228 | |
| 1470 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin SA aa 21-141 Accession # P09228 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Cystatin SA in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant human (rh) Cystatin S and SN and no cross-reactivity with rhCystatin A, B, C, D, E/M, rhFetuin A, B, and rhHPRG (Histidine-Proline-Rich Glycoprotein). |
| 202332 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CST2, CSTSA, cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
| CST2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title