missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin S Antibody, R&D Systems™
Goat Polyclonal Antibody
£418.32
Specifications
Antigen | Cystatin S |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Cystatin S Polyclonal specifically detects Cystatin S in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cystatin S | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CST4, cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1 | |
CST4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P01036 | |
1472 | |
Mouse myeloma cell line NS0-derived recombinant human Cystatin S Ser21-Ala141 Accession # P01036 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Cystatin S in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) Cystatin SA and rhCystatin SN is observed and 5% cross-reactivity with rhCystatin C and rhCystatin D is observed, and less than 1% cross-reactivity with rhCystatin A, rhCystatin B, rhCystatin E/M, rhFetuin A, and rhFetuin B is observed. |
For Research Use Only